missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-hydroxy-4-methoxybenzoate, 98%

Catalog No. AAL0385509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0385509 10 g
AAL0385518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0385509 Supplier Thermo Scientific Chemicals Supplier No. L0385509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5446-02-6 | C9H10O4 | 182.18 g/mol

Methyl 2-hydroxy-4-methoxybenzoate is used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-4-methoxybenzoate is used in the enantioselective synthesis of (+)-coriandrone A and B, two bioactive natural products.

Solubility
Soluble in acetone. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5446-02-6
MDL Number MFCD00008424

Specifications

Melting Point 47°C to 51°C
Boiling Point 134°C (7 mmHg)
Molecular Formula C9H10O4
Beilstein 2691644
Synonym methyl 4-methoxysalicylate, 2-hydroxy-4-methoxy-benzoic acid methyl ester, methyl-4-methoxysalicylate, benzoic acid, 2-hydroxy-4-methoxy-, methyl ester, 2-hydroxy-4-methoxybenzoic acid methyl ester, 4-methoxysalicylic acid methyl ester, acmc-2097hp, methyl 2-hydroxy-p-anisate, ksc494a7p, 2-methoxycarbonyl-5-methoxyphenol
Solubility Information Soluble in acetone. Insoluble in water.
InChI Key ZICRWXFGZCVTBZ-UHFFFAOYSA-N
SMILES COC1=CC(=C(C=C1)C(=O)OC)O
IUPAC Name methyl 2-hydroxy-4-methoxybenzoate
Quantity 10 g
Molecular Weight (g/mol) 182.18
PubChem CID 79523
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material Methyl 2-hydroxy-4-methoxybenzoate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 226-657-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only