Learn More
Methyl 2-furoate, 98+%
CAS: 611-13-2 | C6H6O3 | 126.111 g/mol
Supplier: Thermo Scientific Chemicals A1400936
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl 2-furoate | |
| 1.179 | |
| 73°C (163°F) | |
| 1.487 | |
| 500 g | |
| 111110 | |
| methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester | |
| COC(=O)C1=CC=CO1 | |
| 126.111 | |
| 126.11 |
| 611-13-2 | |
| 180°C to 181°C | |
| C6H6O3 | |
| MFCD00003236 | |
| UN2810 | |
| 14,4307 | |
| HDJLSECJEQSPKW-UHFFFAOYSA-N | |
| methyl furan-2-carboxylate | |
| 11902 | |
| ≥98% |
Chemical Identifiers
| 611-13-2 | |
| 126.111 | |
| HDJLSECJEQSPKW-UHFFFAOYSA-N | |
| 11902 | |
| COC(=O)C1=CC=CO1 |
| C6H6O3 | |
| MFCD00003236 | |
| methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester | |
| methyl furan-2-carboxylate |
Safety and Handling
GHS H Statement
H301-H227-H312-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H301-H312-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 210-254-6
RTECSNumber : LV1950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only