missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-furoate, 98+%

Catalog No. AAA1400914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1400914 25 g
AAA1400922 100 g
AAA1400936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1400914 Supplier Thermo Scientific Chemicals Supplier No. A1400914

CAS: 611-13-2 | C6H6O3 | 126.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-13-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00003236
InChI Key HDJLSECJEQSPKW-UHFFFAOYSA-N
Synonym methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester
PubChem CID 11902
IUPAC Name methyl furan-2-carboxylate
SMILES COC(=O)C1=CC=CO1

Specifications

Density 1.179
Boiling Point 180°C to 181°C
Flash Point 73°C (163°F)
Refractive Index 1.487
Quantity 25 g
UN Number UN2810
Beilstein 111110
Merck Index 14,4307
Formula Weight 126.11
Percent Purity ≥98%
Chemical Name or Material Methyl 2-furoate
Show More Show Less
Hazard Category H227-H301-H312-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227-H312-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 210-254-6
RTECSNumber LV1950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only