missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Furancarboxylate 99.0+%, TCI America™
Supplier: TCI America F0075250ML
Specifications
| Methyl 2-Furancarboxylate | |
| Yellow | |
| C6H6O3 | |
| 250 mL | |
| methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester | |
| COC(=O)C1=CC=CO1 | |
| 126.111 | |
| 126.11 | |
| Liquid |
| 611-13-2 | |
| 181°C | |
| MFCD00003236 | |
| 2810 | |
| HDJLSECJEQSPKW-UHFFFAOYSA-N | |
| methyl furan-2-carboxylate | |
| 11902 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 611-13-2 | |
| 126.111 | |
| HDJLSECJEQSPKW-UHFFFAOYSA-N | |
| 11902 | |
| COC(=O)C1=CC=CO1 |
| C6H6O3 | |
| MFCD00003236 | |
| methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester | |
| methyl furan-2-carboxylate |
Safety and Handling
EINECSNumber : (5)-0046
RTECSNumber : LV1950000
TSCA : Yes