missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Furancarboxylate 99.0+%, TCI America™

Catalog No. F007525ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F007525ML 25 mL
F0075250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F007525ML Supplier TCI America Supplier No. F007525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 611-13-2
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00003236
InChI Key HDJLSECJEQSPKW-UHFFFAOYSA-N
Synonym methyl 2-furoate, methyl pyromucate, methyl 2-furancarboxylate, methyl furoate, 2-furancarboxylic acid, methyl ester, 2-methoxycarbonyl furan, pyromucic acid methyl ester, methyl 2-furylcarboxylate, 2-furoic acid, methyl ester, furoic acid, methyl ester
PubChem CID 11902
IUPAC Name methyl furan-2-carboxylate
SMILES COC(=O)C1=CC=CO1

Specifications

Color Yellow
Boiling Point 181°C
Quantity 25 mL
UN Number 2810
Formula Weight 126.11
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Furancarboxylate
EINECSNumber (5)-0046
RTECSNumber LV1950000
TSCA Yes