missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Fluoro-2-methylpropionate 95.0+%, TCI America™
$194.44 - $567.92
Chemical Identifiers
| CAS | 338-76-1 |
|---|---|
| Molecular Formula | C5H9FO2 |
| Molecular Weight (g/mol) | 120.123 |
| MDL Number | MFCD03265444 |
| InChI Key | OEGBOFOVYSOERL-UHFFFAOYSA-N |
| Synonym | 2-Fluoro-2-methylpropionic Acid Methyl Ester |
| PubChem CID | 9793744 |
| IUPAC Name | methyl 2-fluoro-2-methylpropanoate |
| SMILES | CC(C)(C(=O)OC)F |
Chemical Identifiers
| 338-76-1 | |
| 120.123 | |
| OEGBOFOVYSOERL-UHFFFAOYSA-N | |
| 9793744 | |
| CC(C)(C(=O)OC)F |
| C5H9FO2 | |
| MFCD03265444 | |
| 2-Fluoro-2-methylpropionic Acid Methyl Ester | |
| methyl 2-fluoro-2-methylpropanoate |
Specifications
| 338-76-1 | |
| 109°C | |
| MFCD03265444 | |
| 2924 | |
| OEGBOFOVYSOERL-UHFFFAOYSA-N | |
| methyl 2-fluoro-2-methylpropanoate | |
| 9793744 | |
| ≥95.0% (GC) | |
| Methyl 2-Fluoro-2-methylpropionate |
| Colorless | |
| C5H9FO2 | |
| 5 g | |
| 2-Fluoro-2-methylpropionic Acid Methyl Ester | |
| CC(C)(C(=O)OC)F | |
| 120.123 | |
| 120.12 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3792
TSCA : No