missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Fluoro-2-methylpropionate 95.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 109°C |
| Quantity | 5 g |
| UN Number | 2924 |
| Formula Weight | 120.12 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Methyl 2-Fluoro-2-methylpropionate |
Safety and Handling
| EINECSNumber | (2)-3792 |
| TSCA | No |