missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Méthyl 2-chloropropionate 95,0+%, TCI America™
Spécifications
Spécifications
| Color | Incolore |
| Boiling Point | 132°C |
| Quantity | 500 mL |
| UN Number | 2933 |
| Formula Weight | 122.55 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | Methyl 2-Chloropropionate |
Sécurité et manipulation
| EINECSNumber | (2)-3084 |
| RTECSNumber | UE9100000 |
| TSCA | Yes |