missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-chloropropionate, 97%

Catalog No. AAA1854130
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1854130 250 g
AAA185410B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1854130 Supplier Thermo Scientific Chemicals Supplier No. A1854130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17639-93-9 | C4H7ClO2 | 122.55 g/mol

Methyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-chloropropionate is used as initiator during the synthesis of acrylamide homopolymers and acrylamide-N-isopropylacrylamide block copolymers. It is also used to produce 2-thiophen-2-yl-propionic acid methyl ester.

Solubility
Insoluble in water

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 17639-93-9
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.55
MDL Number MFCD00000863
InChI Key JLEJCNOTNLZCHQ-UHFFFAOYNA-N
Synonym methyl 2-chloropropionate, propanoic acid, 2-chloro-, methyl ester, 2-chloropropionic acid methyl ester, propionic acid, 2-chloro-, methyl ester, methyl alpha-chloropropionate, methyl .alpha.-chloropropionate, .alpha.-chloropropionic acid methyl ester, propanoic acid, 2-chloro-, methyl ester, s, ch3chclc o och3, acmc-1bl6f
PubChem CID 28659
IUPAC Name methyl 2-chloropropanoate
SMILES COC(=O)C(C)Cl

Specifications

Melting Point <-10°C
Density 1.14
Boiling Point 132°C to 133°C
Flash Point 38°C (100°F)
Refractive Index 1.417
Quantity 250 g
UN Number UN2933
Beilstein 1421535
Solubility Information Insoluble in water
Formula Weight 122.55
Percent Purity 97%
Chemical Name or Material Methyl 2-chloropropionate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYL 2-CHLOROPROPIONATE
EINECSNumber 241-624-5
RTECSNumber UE9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only