missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Chloronicotinate 98.0+%, TCI America™

Catalog No. M21655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21655G 5 g
M216525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21655G Supplier TCI America Supplier No. M21655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 40134-18-7 | C7H6ClNO2 | 171.58 g/mol

Chemical Identifiers

CAS 40134-18-7
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD01318629
InChI Key MYGAJZBZLONIBZ-UHFFFAOYSA-N
PubChem CID 819931
IUPAC Name methyl 2-chloropyridine-3-carboxylate
SMILES COC(=O)C1=CC=CN=C1Cl

Specifications

Boiling Point 70°C
Quantity 5 g
Synonym methyl 2-chloronicotinate, 2-chloronicotinic acid methyl ester, 2-chloro-3-pyridinecarboxylic acid methyl ester, methyl2-chloronicotinate, methyl 2-chloro-3-pyridinecarboxylate, 3-pyridinecarboxylic acid, 2-chloro-, methyl ester, 2-chloro-nicotinic acid methyl ester, methyl chloronicotinate, pubchem15917, acmc-1am6z
Formula Weight 171.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Chloronicotinate