missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Chloroisonicotinate 98.0+%, TCI America™

Catalog No. M21645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21645G 5 g
M216425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21645G Supplier TCI America Supplier No. M21645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58481-11-1
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD01765409
InChI Key KKOUHTMLFUAAGG-UHFFFAOYSA-N
Synonym 2-Chloroisonicotinic Acid Methyl Ester, Methyl 2-Chloro-4-pyridinecarboxylate, 2-Chloro-4-pyridinecarboxylic Acid Methyl Ester
PubChem CID 2736842
IUPAC Name methyl 2-chloropyridine-4-carboxylate
SMILES COC(=O)C1=CC(=NC=C1)Cl

Specifications

Melting Point 34°C
Color Yellow
Boiling Point 70°C
Quantity 5 g
Formula Weight 171.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Methyl 2-Chloroisonicotinate
TSCA No
Recommended Storage Refrigerator