missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Chloroacetoacetate 95.0+%, TCI America™
$139.81 - $827.63
Chemical Identifiers
| CAS | 4755-81-1 |
|---|---|
| Molecular Formula | C5H7ClO3 |
| Molecular Weight (g/mol) | 150.558 |
| MDL Number | MFCD00008757 |
| InChI Key | GYQRIAVRKLRQKP-UHFFFAOYSA-N |
| PubChem CID | 107332 |
| IUPAC Name | methyl 2-chloro-3-oxobutanoate |
| SMILES | CC(=O)C(C(=O)OC)Cl |
Chemical Identifiers
| 4755-81-1 | |
| 150.558 | |
| GYQRIAVRKLRQKP-UHFFFAOYSA-N | |
| methyl 2-chloro-3-oxobutanoate |
| C5H7ClO3 | |
| MFCD00008757 | |
| 107332 | |
| CC(=O)C(C(=O)OC)Cl |
Specifications
| 4755-81-1 | |
| 86°C | |
| MFCD00008757 | |
| 1760 | |
| GYQRIAVRKLRQKP-UHFFFAOYSA-N | |
| methyl 2-chloro-3-oxobutanoate | |
| 107332 | |
| ≥95.0% (GC) | |
| Methyl 2-Chloroacetoacetate |
| Yellow | |
| C5H7ClO3 | |
| 25 g | |
| methyl 2-chloroacetoacetate, butanoic acid, 2-chloro-3-oxo-, methyl ester, 2-chloroacetoacetic acid methyl ester, 2-chloro-3-oxo-butyric acid methyl ester, methyl2-chloroacetoacetate, acmc-209k9v, methyl-2-chloroacetoacetate, methyl-alpha-chloroacetoacetate, 2-chloroacetoacetate methyl ester, gyqriavrklrqkp-uhfffaoysa | |
| CC(=O)C(C(=O)OC)Cl | |
| 150.558 | |
| 150.56 | |
| Liquid |
Safety and Handling
TSCA : Yes