missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Chloroacetoacetate 95.0+%, TCI America™

Catalog No. C121925G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C121925G 25 g
C1219250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C121925G Supplier TCI America Supplier No. C121925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4755-81-1
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.558
MDL Number MFCD00008757
InChI Key GYQRIAVRKLRQKP-UHFFFAOYSA-N
PubChem CID 107332
IUPAC Name methyl 2-chloro-3-oxobutanoate
SMILES CC(=O)C(C(=O)OC)Cl

Specifications

Color Yellow
Boiling Point 86°C
Quantity 25 g
UN Number 1760
Synonym methyl 2-chloroacetoacetate, butanoic acid, 2-chloro-3-oxo-, methyl ester, 2-chloroacetoacetic acid methyl ester, 2-chloro-3-oxo-butyric acid methyl ester, methyl2-chloroacetoacetate, acmc-209k9v, methyl-2-chloroacetoacetate, methyl-alpha-chloroacetoacetate, 2-chloroacetoacetate methyl ester, gyqriavrklrqkp-uhfffaoysa
Formula Weight 150.56
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Chloroacetoacetate