missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Bromopropionate 98.0+%, TCI America™
Supplier: TCI America B1460500G
Specifications
| Methyl 2-Bromopropionate | |
| Yellow | |
| C4H7BrO2 | |
| 500 g | |
| methyl 2-bromopropionate, propanoic acid, 2-bromo-, methyl ester, 2-bromopropionic acid methyl ester, methyl dl-2-bromopropionate, methyl2-bromopropionate, methyl alpha-bromopropionate, methyl .alpha.-bromopropionate, methyl-2-bromopropanoate, methyl 2-bromo-propionate, propionic acid, 2-bromo-, methyl ester | |
| CC(C(=O)OC)Br | |
| 167.002 | |
| 167.00 | |
| Liquid |
| 5445-17-0 | |
| 144°C | |
| MFCD00000143 | |
| 2920 | |
| ACEONLNNWKIPTM-UHFFFAOYSA-N | |
| methyl 2-bromopropanoate | |
| 95576 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 5445-17-0 | |
| 167.002 | |
| ACEONLNNWKIPTM-UHFFFAOYSA-N | |
| methyl 2-bromopropanoate |
| C4H7BrO2 | |
| MFCD00000143 | |
| 95576 | |
| CC(C(=O)OC)Br |
Safety and Handling
EINECSNumber : (2)-1179
TSCA : Yes