missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(Bromomethyl)acrylate 97.0+%, TCI America™

Catalog No. M20675G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20675G 5 g
M20671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20675G Supplier TCI America Supplier No. M20675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4224-69-5
Molecular Formula C5H7BrO2
Molecular Weight (g/mol) 179.01
MDL Number MFCD00011697
InChI Key CFTUQSLVERGMHL-UHFFFAOYSA-N
Synonym methyl 2-bromomethyl acrylate, methyl 2-bromomethyl prop-2-enoate, methyl 2-bromomethyl-2-propenoate, methyl alpha-bromomethyl acrylate, 2-bromomethyl methyl acrylate, 2-propenoic acid, 2-bromomethyl-, methyl ester, 2-bromomethyl acrylic acid methyl ester, acrylic acid, 2-bromomethyl-, methyl ester, zlchem 195, pubchem17371
PubChem CID 521093
IUPAC Name methyl 2-(bromomethyl)prop-2-enoate
SMILES COC(=O)C(=C)CBr

Specifications

Color Yellow
Boiling Point 42°C
Quantity 5 g
Formula Weight 179.01
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-(Bromomethyl)acrylate
RTECSNumber AS4900000
TSCA No
Recommended Storage Refrigerator