missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-(bromomethyl)acrylate, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown to Colorless |
| Density | 1.4890g/mL |
| Boiling Point | 35.0°C to 37.0°C (13.0 mmHg) |
| Flash Point | 78°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4870 to 1.4890 |
| Quantity | 1 g |
| Specific Gravity | 1.489 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes serious eye damage. Causes skin irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. |
RUO – Research Use Only