missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-Bromobutyrate 97.0+%, TCI America™

Catalog No. B1462500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1462500G 500 g
B146225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1462500G Supplier TCI America Supplier No. B1462500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3196-15-4
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.029
MDL Number MFCD00009666
InChI Key UFQQDNMQADCHGH-UHFFFAOYSA-N
Synonym methyl 2-bromobutyrate, 2-bromobutyric acid methyl ester, butanoic acid, 2-bromo-, methyl ester, methyl-2-bromobutyrate, alpha-bromobutyric acid methyl ester, methyl bromobutyrate, methyl-2-bromobutanoate, pubchem20191, methylaethylbromessigester, methyl alpha-bromobutyrate
PubChem CID 95577
IUPAC Name methyl 2-bromobutanoate
SMILES CCC(C(=O)OC)Br

Specifications

Color Yellow
Boiling Point 172°C
Quantity 500 g
UN Number 3265
Formula Weight 181.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2-Bromobutyrate
EINECSNumber (2)-1179
TSCA Yes