missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-bromo-3-furoate, 97%, Thermo Scientific™

Catalog No. p-3586845
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC64722CB 250mg Amber Glass Bottle
CC64722DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC64722CB Supplier Thermo Scientific Chemicals Supplier No. CC64722CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 197846-06-3
Molecular Formula C6H5BrO3
Molecular Weight (g/mol) 205.01
MDL Number MFCD09879944
InChI Key XXUUZVIMSVBWIQ-UHFFFAOYSA-N
Synonym methyl 2-bromo-3-furoate, 3-furancarboxylic acid, 2-bromo-, methyl ester, 2-bromofuran-3-carboxylic acid methyl ester, 3-furancarboxylic acid,2-bromo-, methyl ester
PubChem CID 10845885
IUPAC Name methyl 2-bromofuran-3-carboxylate
SMILES COC(=O)C1=C(Br)OC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 205.01
Percent Purity 97%
Chemical Name or Material Methyl 2-bromo-3-furoate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.