missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-aminothiophene-3-carboxylate, 97%

Catalog No. p-7036978
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5548203 1 g
AAH5548206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5548203 Supplier Thermo Scientific Chemicals Supplier No. H5548203

CAS: 4651-81-4 | C6H7NO2S | 157.19 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store at room temperature. Avoid strong oxidizing agents.

Chemical Identifiers

CAS 4651-81-4
Molecular Formula C6H7NO2S
Molecular Weight (g/mol) 157.19
MDL Number MFCD00159547
InChI Key DGGJQLCAYQCPDD-UHFFFAOYSA-N
Synonym methyl 2-amino-3-thenoate, methyl 2-amino-3-thiophenecarboxylate, 2-amino-3-methoxycarbonylthiophene, methyl2-aminothiophene-3-carboxylate, 2-amino-thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylic acid, 2-amino-, methyl ester, zlchem 402, pubchem5551, maybridge1_003714, 2-amino-thiophene-3-carboxylicacidmethylester
PubChem CID 78381
IUPAC Name methyl 2-aminothiophene-3-carboxylate
SMILES COC(=O)C1=C(N)SC=C1

Specifications

Melting Point 76°C to 81°C
Quantity 1 g
Formula Weight 157.19
Percent Purity 97%
Chemical Name or Material Methyl 2-aminothiophene-3-carboxylate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 225-084-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only