missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-3-bromo-5-methoxybenzoate, 96%, Thermo Scientific™

Chemical Identifiers

CAS 1378874-22-6
Molecular Formula C9H10BrNO3
Molecular Weight (g/mol) 260.087
MDL Number MFCD20527215
InChI Key WPRSGBOUSTUNEC-UHFFFAOYSA-N
Synonym 2-amino-3-bromo-5-methoxybenzoic acid methyl ester
PubChem CID 73995991
IUPAC Name methyl 2-amino-3-bromo-5-methoxybenzoate
SMILES COC1=CC(=C(C(=C1)C(=O)OC)N)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH5484403
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

1378874-22-6
260.087
WPRSGBOUSTUNEC-UHFFFAOYSA-N
73995991
COC1=CC(=C(C(=C1)C(=O)OC)N)Br
C9H10BrNO3
MFCD20527215
2-amino-3-bromo-5-methoxybenzoic acid methyl ester
methyl 2-amino-3-bromo-5-methoxybenzoate

Specifications

1378874-22-6
MFCD20527215
WPRSGBOUSTUNEC-UHFFFAOYSA-N
methyl 2-amino-3-bromo-5-methoxybenzoate
73995991
96%
C9H10BrNO3
2-amino-3-bromo-5-methoxybenzoic acid methyl ester
COC1=CC(=C(C(=C1)C(=O)OC)N)Br
260.087
260.09
Methyl 2-amino-3-bromo-5-methoxybenzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only