missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,6-dichlorophenylacetate, 99%

Catalog No. p-162058
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC351070250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC351070250 Supplier Thermo Scientific Chemicals Supplier No. 351070250

CAS: 54551-83-6 | C9H8Cl2O2 | 219.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54551-83-6
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.07
MDL Number MFCD00191640
InChI Key FCWRUYPZZJPCCG-UHFFFAOYSA-N
Synonym methyl 2,6-dichlorophenylacetate, methyl 2-2,6-dichlorophenyl acetate, 2,6-dichlorophenylacetic acid methyl ester, benzeneacetic acid,2,6-dichloro-, methyl ester, benzeneacetic acid, 2,6-dichloro-, methyl ester, acmc-209lhf, methyl2,6-dichlorophenylacetate, guanfacine methyl ester impurity, methyl 2,6-dichlorophenyl acetate, 2,6-dichloro-phenylacetic acid methyl ester
PubChem CID 2734107
IUPAC Name methyl 2-(2,6-dichlorophenyl)acetate
SMILES COC(=O)CC1=C(C=CC=C1Cl)Cl

Specifications

Color Colorless
Density 1.3300g/mL
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5400 to 1.542
Quantity 25 g
Specific Gravity 1.33
Formula Weight 219.07
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl 2, 6-dichlorophenylacetate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 259-221-8

RUO – Research Use Only