missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2,6-Dichloronicotinate 98.0+%, TCI America™
$80.16 - $242.33
Chemical Identifiers
| CAS | 65515-28-8 |
|---|---|
| Molecular Formula | C7H5Cl2NO2 |
| Molecular Weight (g/mol) | 206.022 |
| MDL Number | MFCD07369794 |
| InChI Key | IFVVGOJYWCHRCT-UHFFFAOYSA-N |
| Synonym | methyl 2,6-dichloronicotinate, 2,6-dichloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2,6-dichloro-, methyl ester, acmc-20akiy, methyl 2,6-dichloro-nicotinate, 2,6-dichloro-nicotinic acid methyl ester, methyl-2,6-dichloropyridine-3-carboxylate, methyl-2,6-dichloro-pyridine-3-carboxylate, methyl 2,6-dichloropyridine-3-carboxylate, 2,6-dichloropyridine-3-carboxylic acid methyl ester |
| PubChem CID | 11106516 |
| IUPAC Name | methyl 2,6-dichloropyridine-3-carboxylate |
| SMILES | COC(=O)C1=C(N=C(C=C1)Cl)Cl |
Chemical Identifiers
| 65515-28-8 | |
| 206.022 | |
| IFVVGOJYWCHRCT-UHFFFAOYSA-N | |
| 11106516 | |
| COC(=O)C1=C(N=C(C=C1)Cl)Cl |
| C7H5Cl2NO2 | |
| MFCD07369794 | |
| methyl 2,6-dichloronicotinate, 2,6-dichloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2,6-dichloro-, methyl ester, acmc-20akiy, methyl 2,6-dichloro-nicotinate, 2,6-dichloro-nicotinic acid methyl ester, methyl-2,6-dichloropyridine-3-carboxylate, methyl-2,6-dichloro-pyridine-3-carboxylate, methyl 2,6-dichloropyridine-3-carboxylate, 2,6-dichloropyridine-3-carboxylic acid methyl ester | |
| methyl 2,6-dichloropyridine-3-carboxylate |
Specifications
| 65515-28-8 | |
| White-Yellow | |
| MFCD07369794 | |
| methyl 2,6-dichloronicotinate, 2,6-dichloronicotinic acid methyl ester, 3-pyridinecarboxylic acid, 2,6-dichloro-, methyl ester, acmc-20akiy, methyl 2,6-dichloro-nicotinate, 2,6-dichloro-nicotinic acid methyl ester, methyl-2,6-dichloropyridine-3-carboxylate, methyl-2,6-dichloro-pyridine-3-carboxylate, methyl 2,6-dichloropyridine-3-carboxylate, 2,6-dichloropyridine-3-carboxylic acid methyl ester | |
| COC(=O)C1=C(N=C(C=C1)Cl)Cl | |
| 206.022 | |
| 206.02 | |
| Crystalline Powder |
| 59°C | |
| C7H5Cl2NO2 | |
| 1 g | |
| IFVVGOJYWCHRCT-UHFFFAOYSA-N | |
| methyl 2,6-dichloropyridine-3-carboxylate | |
| 11106516 | |
| ≥98.0% (GC) | |
| Methyl 2,6-Dichloronicotinate |
Safety and Handling
TSCA : No