missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Methyl 2,6-dichlorobenzoate, 97%

Catalog No. AAB2491206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2491206 5 g
AAB2491203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2491206 Supplier Thermo Scientific™ Supplier No. B2491206
Only null left

CAS: 14920-87-7 | C8H6Cl2O2 | 205.03 g/mol

Chemical Identifiers

CAS 14920-87-7
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.03
MDL Number MFCD01313701
InChI Key HEYGSGDIWJDORA-UHFFFAOYSA-N
Synonym 2,6-Dichlorobenzoic acid methyl ester;
IUPAC Name methyl 2,6-dichlorobenzoate
SMILES COC(=O)C1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 25-30°
Boiling Point 250°
Quantity 5 g
Formula Weight 205.04
Physical Form Solid
ShelfLife 5 Years