missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,5-dichlorothiophene-3-carboxylate, 98%

Catalog No. p-7044762
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0811803 1 g
AAL0811806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0811803 Supplier Thermo Scientific Chemicals Supplier No. L0811803

CAS: 145129-54-0 | C6H4Cl2O2S | 211.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 145129-54-0
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.056
MDL Number MFCD00051763
InChI Key VCXPHMKCDRPIDG-UHFFFAOYSA-N
Synonym acmc-1c2sm, methyl 2,5-dichloro-3-thiophenecarboxylate, methyl-2,5-dichlorothiophene-3-carboxylate, methyl 2,5-dichloro-3-thiophenecarboxylate #, methyl 2,5-bis chloranyl thiophene-3-carboxylate, 2,5-dichloro-3-thiophenecarboxylic acid methyl ester, 3-thiophenecarboxylic acid, 2,5-dichloro-, methyl ester, 3-thiophenecarboxylicacid, 2,5-dichloro-, methyl ester
PubChem CID 518961
IUPAC Name methyl 2,5-dichlorothiophene-3-carboxylate
SMILES COC(=O)C1=C(SC(=C1)Cl)Cl

Specifications

Boiling Point 74°C to 75°C (0.6 mmHg)
Refractive Index 1.563
Quantity 1 g
Formula Weight 211.07
Percent Purity 98%
Chemical Name or Material Methyl 2,5-dichlorothiophene-3-carboxylate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only