missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,4,6-trihydroxybenzoate, 98%

Catalog No. p-7082054
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1230506 5 g
AAA1230514 25 g
AAA1230522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1230506 Supplier Thermo Scientific Chemicals Supplier No. A1230506

CAS: 3147-39-5 | C8H8O5 | 184.147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3147-39-5
Molecular Formula C8H8O5
Molecular Weight (g/mol) 184.147
MDL Number MFCD00013969
InChI Key AQDIJIAUYXOCGX-UHFFFAOYSA-N
PubChem CID 76600
IUPAC Name methyl 2,4,6-trihydroxybenzoate
SMILES COC(=O)C1=C(C=C(C=C1O)O)O

Specifications

Melting Point 175°C to 179°C
Quantity 5 g
Beilstein 2837819
Synonym benzoic acid, 2,4,6-trihydroxy-, methyl ester, 2,4,6-trihydroxybenzoic acid methyl ester, acmc-20akj4, benzoic acid,2,4,6-trihydroxy-, methyl ester
Formula Weight 184.15
Percent Purity 98%
Chemical Name or Material Methyl 2,4,6-trihydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-566-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only