missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2,3-O-isopropylidene-beta-D-ribofuranoside, 98%
CAS: 4099-85-8 | C9H16O5 | 204.22 g/mol
$96.93 - $370.71
Chemical Identifiers
| CAS | 4099-85-8 |
|---|---|
| Molecular Formula | C9H16O5 |
| Molecular Weight (g/mol) | 204.22 |
| MDL Number | MFCD00270040 |
| InChI Key | DXBHDBLZPXQALN-UHFFFAOYNA-N |
| Synonym | methyl 2,3-o-isopropylidene-beta-d-ribofuranoside, 3ar,4r,6r,6ar-6-methoxy-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, methyl-2,3-o-isopropylidene-beta-d-ribofuranoside, methyl 2,3-o-isopropylidene-b-d-ribofuranoside, methyl 2,3-o-1-methylethylidene-, a-d-ribofuranoside, 3ar,4r,6r,6ar-6-methoxy-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, pubchem10741, methyl 2,3-o-isopropylidene-, methyl 2-o,3-o-isopropylidene-beta-d-ribofuranoside |
| PubChem CID | 96666 |
| IUPAC Name | {6-methoxy-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl}methanol |
| SMILES | COC1OC(CO)C2OC(C)(C)OC12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6629603
|
Thermo Scientific Chemicals
J6629603 |
1 g |
Each for $96.93
|
|
|||||
|
AAJ6629606
|
Thermo Scientific Chemicals
J6629606 |
5 g |
Each for $370.71
|
|
|||||
Description
Methyl 2,3-O-isopropylidene-beta-D-ribofuranoside is used for the manufacture of fine chemicals, and Industrial use resulting in manufacture of intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 2,3-O-isopropylidene-beta-D-ribofuranoside is used for the manufacture of fine chemicals, and Industrial use resulting in manufacture of intermediates.
Solubility
Soluble in chloroform, dichloromethane, ethanol, ethyl acetate, and methanol
Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 4099-85-8 | |
| 204.22 | |
| DXBHDBLZPXQALN-UHFFFAOYNA-N | |
| 96666 | |
| COC1OC(CO)C2OC(C)(C)OC12 |
| C9H16O5 | |
| MFCD00270040 | |
| methyl 2,3-o-isopropylidene-beta-d-ribofuranoside, 3ar,4r,6r,6ar-6-methoxy-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, methyl-2,3-o-isopropylidene-beta-d-ribofuranoside, methyl 2,3-o-isopropylidene-b-d-ribofuranoside, methyl 2,3-o-1-methylethylidene-, a-d-ribofuranoside, 3ar,4r,6r,6ar-6-methoxy-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, pubchem10741, methyl 2,3-o-isopropylidene-, methyl 2-o,3-o-isopropylidene-beta-d-ribofuranoside | |
| {6-methoxy-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl}methanol |
Specifications
| 4099-85-8 | |
| 1 g | |
| C9H16O5 | |
| methyl 2,3-o-isopropylidene-beta-d-ribofuranoside, 3ar,4r,6r,6ar-6-methoxy-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, methyl-2,3-o-isopropylidene-beta-d-ribofuranoside, methyl 2,3-o-isopropylidene-b-d-ribofuranoside, methyl 2,3-o-1-methylethylidene-, a-d-ribofuranoside, 3ar,4r,6r,6ar-6-methoxy-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, pubchem10741, methyl 2,3-o-isopropylidene-, methyl 2-o,3-o-isopropylidene-beta-d-ribofuranoside | |
| DXBHDBLZPXQALN-UHFFFAOYNA-N | |
| {6-methoxy-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl}methanol | |
| 96666 | |
| 98% |
| Yellow | |
| Oil | |
| MFCD00270040 | |
| Soluble in chloroform, dichloromethane, ethanol, ethyl acetate, and methanol | |
| COC1OC(CO)C2OC(C)(C)OC12 | |
| 204.22 | |
| 204.22 | |
| Methyl 2,3-O-isopropylidene-beta-D-ribofuranoside |
Safety and Handling
EINECSNumber : 223-865-8
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only