missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,3-O-isopropylidene-beta-D-ribofuranoside, 98%

Catalog No. AAJ6629606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6629606 5 g
AAJ6629603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6629606 Supplier Thermo Scientific Chemicals Supplier No. J6629606

CAS: 4099-85-8 | C9H16O5 | 204.22 g/mol

Methyl 2,3-O-isopropylidene-beta-D-ribofuranoside is used for the manufacture of fine chemicals, and Industrial use resulting in manufacture of intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,3-O-isopropylidene-beta-D-ribofuranoside is used for the manufacture of fine chemicals, and Industrial use resulting in manufacture of intermediates.

Solubility
Soluble in chloroform, dichloromethane, ethanol, ethyl acetate, and methanol

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 4099-85-8
Molecular Formula C9H16O5
Molecular Weight (g/mol) 204.22
MDL Number MFCD00270040
InChI Key DXBHDBLZPXQALN-UHFFFAOYNA-N
Synonym methyl 2,3-o-isopropylidene-beta-d-ribofuranoside, 3ar,4r,6r,6ar-6-methoxy-2,2-dimethyltetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, methyl-2,3-o-isopropylidene-beta-d-ribofuranoside, methyl 2,3-o-isopropylidene-b-d-ribofuranoside, methyl 2,3-o-1-methylethylidene-, a-d-ribofuranoside, 3ar,4r,6r,6ar-6-methoxy-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl methanol, pubchem10741, methyl 2,3-o-isopropylidene-, methyl 2-o,3-o-isopropylidene-beta-d-ribofuranoside
PubChem CID 96666
IUPAC Name {6-methoxy-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl}methanol
SMILES COC1OC(CO)C2OC(C)(C)OC12

Specifications

Color Yellow
Quantity 5 g
Physical Form Oil
Solubility Information Soluble in chloroform, dichloromethane, ethanol, ethyl acetate, and methanol
Formula Weight 204.22
Percent Purity 98%
Chemical Name or Material Methyl 2,3-O-isopropylidene-beta-D-ribofuranoside
EINECSNumber 223-865-8
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only