missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,3,4-Tri-O-acetyl-1-thio-beta-L-fucopyranoside 98.0+%, TCI America™

Supplier:  TCI America M16261G

 View more versions of this product

Catalog No. M16261G


Only null left

Description

Description

Specifications

Specifications

Methyl 2,3,4-Tri-O-acetyl-1-thio-beta-L-fucopyranoside
148°C
C13H20O7S
1 g
CC1C(C(C(C(O1)SC)OC(=O)C)OC(=O)C)OC(=O)C
320.356
320.36
Crystalline Powder
84635-54-1
White-Yellow
MFCD00080803
JOLBZRHXWOHZNS-MCNNAKBESA-N
[(2S,3R,4R,5S,6R)-4,5-diacetyloxy-2-methyl-6-methylsulfanyloxan-3-yl] acetate
14131721
≥98.0% (HPLC)

Chemical Identifiers

84635-54-1
320.356
JOLBZRHXWOHZNS-MCNNAKBESA-N
[(2S,3R,4R,5S,6R)-4,5-diacetyloxy-2-methyl-6-methylsulfanyloxan-3-yl] acetate
C13H20O7S
MFCD00080803
14131721
CC1C(C(C(C(O1)SC)OC(=O)C)OC(=O)C)OC(=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions