missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,2-Dimethoxypropionate 96.0+%, TCI America™

Catalog No. D238225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D238225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D238225G Supplier TCI America Supplier No. D238225G
Only null left

Chemical Identifiers

CAS 10076-48-9
Molecular Formula C6H12O4
Molecular Weight (g/mol) 148.16
MDL Number MFCD00048066
InChI Key KHQQDBIXHUJARJ-UHFFFAOYSA-N
Synonym 2,2-Dimethoxypropionic Acid Methyl Ester
PubChem CID 82339
IUPAC Name methyl 2,2-dimethoxypropanoate
SMILES COC(=O)C(C)(OC)OC

Specifications

Color Colorless
Boiling Point 70°C
Quantity 25 g
Formula Weight 148.16
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 2,2-Dimethoxypropionate