missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,2,2-trichloroacetimidate, 98%

Catalog No. AC160931000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160931000 100 g Glass bottle
AC160930250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC160931000 Supplier Thermo Scientific Chemicals Supplier No. 160931000
Only null left

CAS: 2533-69-9 | C3H4Cl3NO | 176.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2533-69-9
Molecular Formula C3H4Cl3NO
Molecular Weight (g/mol) 176.42
MDL Number MFCD00001759
InChI Key OGBINJLTBZWRRB-UHFFFAOYSA-N
Synonym methyl 2,2,2-trichloroacetimidate, methyl trichloroacetimidate, methyl 2,2,2-trichloroethanecarboximidate, ethanimidic acid, 2,2,2-trichloro-, methyl ester, acmc-20aebo, methyltrichloroacetimidate, methyl trichloroacetoimidate, methyl-2,2,2-trichloroacetimidate, methyl 2,2,2-trichloroethanimidoate, trichloroacetimidic acid methyl ester
PubChem CID 100479
IUPAC Name methyl 2,2,2-trichloroethanimidate
SMILES COC(=N)C(Cl)(Cl)Cl

Specifications

Color Colorless to Yellow
Density 1.4200g/mL
Boiling Point 148.0°C to 149.0°C
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4770 to 1.4790
Linear Formula CCl3C(=NH)OCH3
Quantity 100 g
Beilstein 02, 212
Specific Gravity 1.42
Formula Weight 176.43
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 2,2,2-trichloroacetimidate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 219-796-8

RUO – Research Use Only