missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1H-Imidazole-5-carboxylate 97.0+%, TCI America™

$48.46 - $180.81

Chemical Identifiers

CAS 17325-26-7
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00216584
InChI Key DVLGIQNHKLWSRU-UHFFFAOYSA-N
Synonym methyl 4-imidazolecarboxylate, methyl 1h-imidazole-4-carboxylate, methyl imidazole-4-carboxylate, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, methyl-4-imidazolecarboxylate, 1h-imidazole-4-carboxylic acid methyl ester, methyl 3h-imidazole-4-carboxylate, imidazole-4-carboxylic acid, methyl ester, zlchem 863
PubChem CID 565661
IUPAC Name methyl 1H-imidazole-5-carboxylate
SMILES COC(=O)C1=CN=CN1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M22281G
View Documents
TCI America
M22281G
1 g
Each for $48.46
Only null left
 
M22285G
View Documents
TCI America
M22285G
5 g
Each for $180.81
Only null left
 
Description

Description

Specifications

Chemical Identifiers

17325-26-7
126.12
DVLGIQNHKLWSRU-UHFFFAOYSA-N
565661
COC(=O)C1=CN=CN1
C5H6N2O2
MFCD00216584
methyl 4-imidazolecarboxylate, methyl 1h-imidazole-4-carboxylate, methyl imidazole-4-carboxylate, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, methyl-4-imidazolecarboxylate, 1h-imidazole-4-carboxylic acid methyl ester, methyl 3h-imidazole-4-carboxylate, imidazole-4-carboxylic acid, methyl ester, zlchem 863
methyl 1H-imidazole-5-carboxylate

Specifications

17325-26-7
C5H6N2O2
1 g
DVLGIQNHKLWSRU-UHFFFAOYSA-N
methyl 1H-imidazole-5-carboxylate
565661
≥97.0% (GC,T)
Methyl 1H-Imidazole-5-carboxylate
153°C
MFCD00216584
methyl 4-imidazolecarboxylate, methyl 1h-imidazole-4-carboxylate, methyl imidazole-4-carboxylate, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, methyl-4-imidazolecarboxylate, 1h-imidazole-4-carboxylic acid methyl ester, methyl 3h-imidazole-4-carboxylate, imidazole-4-carboxylic acid, methyl ester, zlchem 863
COC(=O)C1=CN=CN1
126.12
126.12
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'