missing translation for 'onlineSavingsMsg'
Learn More

Méthyl 1H-Imidazole-5-carboxylate 97,0+%, TCI America™

Numéro de catalogue. M22285G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M22285G 5 g
M22281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M22285G Fournisseur TCI America Code fournisseur M22285G

Identifiants chimiques

CAS 17325-26-7
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00216584
InChI Key DVLGIQNHKLWSRU-UHFFFAOYSA-N
Synonym methyl 4-imidazolecarboxylate, methyl 1h-imidazole-4-carboxylate, methyl imidazole-4-carboxylate, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, methyl-4-imidazolecarboxylate, 1h-imidazole-4-carboxylic acid methyl ester, methyl 3h-imidazole-4-carboxylate, imidazole-4-carboxylic acid, methyl ester, zlchem 863
PubChem CID 565661
IUPAC Name méthyle 1H-imidazole-5-carboxylate
SMILES COC(=O)C1=CN=CN1

Spécifications

Melting Point 153°C
Quantity 5 g
Formula Weight 126.12
Percent Purity ≥97.0% (GC,T)
Physical Form Poudre cristalline
Chemical Name or Material Methyl 1H-Imidazole-5-carboxylate