missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1H-Imidazole-5-carboxylate 97.0+%, TCI America™

Catalog No. M22281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22281G 1 g
M22285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22281G Supplier TCI America Supplier No. M22281G

Chemical Identifiers

CAS 17325-26-7
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00216584
InChI Key DVLGIQNHKLWSRU-UHFFFAOYSA-N
Synonym methyl 4-imidazolecarboxylate, methyl 1h-imidazole-4-carboxylate, methyl imidazole-4-carboxylate, 1h-imidazole-4-carboxylic acid, methyl ester, 1h-imidazole-5-carboxylic acid methyl ester, methyl-4-imidazolecarboxylate, 1h-imidazole-4-carboxylic acid methyl ester, methyl 3h-imidazole-4-carboxylate, imidazole-4-carboxylic acid, methyl ester, zlchem 863
PubChem CID 565661
IUPAC Name methyl 1H-imidazole-5-carboxylate
SMILES COC(=O)C1=CN=CN1

Specifications

Melting Point 153°C
Quantity 1 g
Formula Weight 126.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1H-Imidazole-5-carboxylate