missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylate 98.0+%, TCI America™

Catalog No. M27231G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27231G 1 g
M27235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27231G Supplier TCI America Supplier No. M27231G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 122684-33-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD04038683
InChI Key LIWFYAVKYUQMRE-UHFFFAOYSA-N
Synonym methyl 1-boc-3-pyrrolidinecarboxylate, 1-tert-butyl 3-methyl pyrrolidine-1,3-dicarboxylate, methyl 1-n-boc-dl-beta-prolinate, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, methyl 1-boc-3-pyrrolidine carboxylate, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, methyl 1-boc-pyrrolidine-3-carboxylate, methyl 1-tert-butoxycarbonyl-3-pyrrolidinecarboxylate, s-1-boc-pyrrolidine-1,3-dicarboxylate, pubchem10748
PubChem CID 4572775
IUPAC Name 1-O-tert-butyl 3-O-methyl pyrrolidine-1,3-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)C(=O)OC

Specifications

Color Yellow
Boiling Point 105°C
Quantity 1 g
Formula Weight 229.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylate