missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate 98.0+%, TCI America™

Catalog No. M29185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29185G 5 g
M291825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29185G Supplier TCI America Supplier No. M29185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 148763-41-1
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD06795926
InChI Key LYYJQMCPEHXOEW-UHFFFAOYSA-N
Synonym Methyl 1-Boc-3-piperidinecarboxylate, 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylic Acid Methyl Ester, 1-Boc-3-piperidinecarboxylic Acid Methyl Ester, 1-tert-Butyl 3-Methyl Piperidine-1,3-dicarboxylate, Piperidine-1,3-dicarboxylic Acid 1-tert-Butyl 3-Met
PubChem CID 23590652
IUPAC Name 1-O-tert-butyl 3-O-methyl piperidine-1,3-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C(=O)OC

Specifications

Melting Point 49°C
Color White-Yellow
Quantity 5 g
Formula Weight 243.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate