missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 1-Methyl-4-piperidinecarboxylate 98.0+%, TCI America™
$48.46 - $137.94
Chemical Identifiers
| CAS | 1690-75-1 |
|---|---|
| Molecular Formula | C8H15NO2 |
| Molecular Weight (g/mol) | 157.213 |
| MDL Number | MFCD06658483 |
| InChI Key | KYAOKPRJTMFBTQ-UHFFFAOYSA-N |
| Synonym | 1-Methyl-4-piperidinecarboxylic Acid Methyl Ester |
| PubChem CID | 3616844 |
| IUPAC Name | methyl 1-methylpiperidine-4-carboxylate |
| SMILES | CN1CCC(CC1)C(=O)OC |
Chemical Identifiers
| 1690-75-1 | |
| 157.213 | |
| KYAOKPRJTMFBTQ-UHFFFAOYSA-N | |
| 3616844 | |
| CN1CCC(CC1)C(=O)OC |
| C8H15NO2 | |
| MFCD06658483 | |
| 1-Methyl-4-piperidinecarboxylic Acid Methyl Ester | |
| methyl 1-methylpiperidine-4-carboxylate |
Specifications
| 1690-75-1 | |
| 202°C | |
| MFCD06658483 | |
| 1-Methyl-4-piperidinecarboxylic Acid Methyl Ester | |
| CN1CCC(CC1)C(=O)OC | |
| 157.213 | |
| 157.21 | |
| Liquid |
| Yellow | |
| C8H15NO2 | |
| 1 g | |
| KYAOKPRJTMFBTQ-UHFFFAOYSA-N | |
| methyl 1-methylpiperidine-4-carboxylate | |
| 3616844 | |
| ≥98.0% (GC,T) | |
| Methyl 1-Methyl-4-piperidinecarboxylate |
Safety and Handling
TSCA : No