missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methyl-1H-imidazole-4-carboxylate, 97%, Thermo Scientific™

$145.24 - $145.24

Chemical Identifiers

CAS 17289-19-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD02179561
InChI Key KZPZTVKOJSKVBV-UHFFFAOYSA-N
Synonym methyl 1-methyl-1h-imidazole-4-carboxylate, 1-methyl-1h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-4-carboxylic acid, 1-methyl-, methyl ester, pubchem9002, acmc-209e5g, methyl1-methyl-1h-imidazole-4-carboxylate, ksc534i0d, methyl 1-methylimidazol-4-carboxylate, methyl 1-methyl-1h-imidazole-carboxylate, 4-methoxycarbonyl-1-methyl-1h-imidazole
PubChem CID 2773510
IUPAC Name methyl 1-methylimidazole-4-carboxylate
SMILES CN1C=C(N=C1)C(=O)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC03622DA
SDS View Documents
Thermo Scientific Chemicals
CC03622DA
1 g Amber Glass Bottle
Each for $145.24
Only null left
 
Description

Description

Specifications

Chemical Identifiers

17289-19-9
140.142
KZPZTVKOJSKVBV-UHFFFAOYSA-N
2773510
CN1C=C(N=C1)C(=O)OC
C6H8N2O2
MFCD02179561
methyl 1-methyl-1h-imidazole-4-carboxylate, 1-methyl-1h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-4-carboxylic acid, 1-methyl-, methyl ester, pubchem9002, acmc-209e5g, methyl1-methyl-1h-imidazole-4-carboxylate, ksc534i0d, methyl 1-methylimidazol-4-carboxylate, methyl 1-methyl-1h-imidazole-carboxylate, 4-methoxycarbonyl-1-methyl-1h-imidazole
methyl 1-methylimidazole-4-carboxylate

Specifications

17289-19-9
100.0
Amber Glass Bottle
MFCD02179561
methyl 1-methyl-1h-imidazole-4-carboxylate, 1-methyl-1h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-4-carboxylic acid, 1-methyl-, methyl ester, pubchem9002, acmc-209e5g, methyl1-methyl-1h-imidazole-4-carboxylate, ksc534i0d, methyl 1-methylimidazol-4-carboxylate, methyl 1-methyl-1h-imidazole-carboxylate, 4-methoxycarbonyl-1-methyl-1h-imidazole
CN1C=C(N=C1)C(=O)OC
140.142
140.14
1-Methyl-1H-imidazole-4-carboxylic acid methyl ester
96.5
97%
C6H8N2O2
1 g
KZPZTVKOJSKVBV-UHFFFAOYSA-N
methyl 1-methylimidazole-4-carboxylate
2773510
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'