missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-methyl-1H-imidazole-4-carboxylate, 97%, Thermo Scientific™

Catalog No. CC03622DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC03622DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC03622DA Supplier Thermo Scientific Chemicals Supplier No. CC03622DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17289-19-9
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD02179561
InChI Key KZPZTVKOJSKVBV-UHFFFAOYSA-N
Synonym methyl 1-methyl-1h-imidazole-4-carboxylate, 1-methyl-1h-imidazole-4-carboxylic acid methyl ester, 1h-imidazole-4-carboxylic acid, 1-methyl-, methyl ester, pubchem9002, acmc-209e5g, methyl1-methyl-1h-imidazole-4-carboxylate, ksc534i0d, methyl 1-methylimidazol-4-carboxylate, methyl 1-methyl-1h-imidazole-carboxylate, 4-methoxycarbonyl-1-methyl-1h-imidazole
PubChem CID 2773510
IUPAC Name methyl 1-methylimidazole-4-carboxylate
SMILES CN1C=C(N=C1)C(=O)OC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-imidazole-4-carboxylic acid methyl ester
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.