missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-cyclopentene-1-carboxylate, 95%

Catalog No. AC397590010
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397590010 1 g Glass bottle
AC397590100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397590010 Supplier Thermo Scientific Chemicals Supplier No. 397590010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25662-28-6 | C7H10O2 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25662-28-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00239506
InChI Key VTYCAXIAUKEGBQ-UHFFFAOYSA-N
Synonym methyl 1-cyclopentene-1-carboxylate, methyl cyclopent-1-enecarboxylate, methyl cyclopent-1-ene-1-carboxylate, cyclopentene-1-carboxylic acid methyl ester, methyl 1-cyclopentenecarboxylate, 1-cyclopentenecarboxylic acid methyl ester, 1-cyclopentene-1-carboxylic acid, methyl ester, methyl1-cyclopentene-1-carboxylate, 1-cyclopentene-1-carboxylic acid methyl ester, pubchem2487
PubChem CID 549129
IUPAC Name methyl cyclopentene-1-carboxylate
SMILES COC(=O)C1=CCCC1

Specifications

Color White to Yellow
Density 1.0310g/mL
Boiling Point 76°C to 78°C (9.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C5H7COOCH3
Quantity 1 g
Specific Gravity 1.031
Formula Weight 126.15
Percent Purity 97%
Physical Form Crystalline Powder or Low Melting Solid
Chemical Name or Material Methyl 1-cyclopentene-1-carboxylate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only