missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 1-Boc-pyrrolidine-3-carboxylate, 97%
CAS: 122684-33-7 | C11H19NO4 | 229.276 g/mol
$206.53 - $674.34
Chemical Identifiers
| CAS | 122684-33-7 |
|---|---|
| Molecular Formula | C11H19NO4 |
| Molecular Weight (g/mol) | 229.276 |
| MDL Number | MFCD04038683 |
| InChI Key | LIWFYAVKYUQMRE-UHFFFAOYSA-N |
| Synonym | methyl 1-boc-3-pyrrolidinecarboxylate, 1-tert-butyl 3-methyl pyrrolidine-1,3-dicarboxylate, methyl 1-n-boc-dl-beta-prolinate, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, methyl 1-boc-3-pyrrolidine carboxylate, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, methyl 1-boc-pyrrolidine-3-carboxylate, methyl 1-tert-butoxycarbonyl-3-pyrrolidinecarboxylate, s-1-boc-pyrrolidine-1,3-dicarboxylate, pubchem10748 |
| PubChem CID | 4572775 |
| IUPAC Name | 1-O-tert-butyl 3-O-methyl pyrrolidine-1,3-dicarboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCC(C1)C(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3340703
|
Thermo Scientific Chemicals
H3340703 |
1 g |
Each for $206.53
|
|
|||||
|
AAH3340706
|
Thermo Scientific Chemicals
H3340706 |
5 g |
Each for $674.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 122684-33-7 | |
| 229.276 | |
| LIWFYAVKYUQMRE-UHFFFAOYSA-N | |
| 4572775 | |
| CC(C)(C)OC(=O)N1CCC(C1)C(=O)OC |
| C11H19NO4 | |
| MFCD04038683 | |
| methyl 1-boc-3-pyrrolidinecarboxylate, 1-tert-butyl 3-methyl pyrrolidine-1,3-dicarboxylate, methyl 1-n-boc-dl-beta-prolinate, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, methyl 1-boc-3-pyrrolidine carboxylate, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, methyl 1-boc-pyrrolidine-3-carboxylate, methyl 1-tert-butoxycarbonyl-3-pyrrolidinecarboxylate, s-1-boc-pyrrolidine-1,3-dicarboxylate, pubchem10748 | |
| 1-O-tert-butyl 3-O-methyl pyrrolidine-1,3-dicarboxylate |
Specifications
| 122684-33-7 | |
| C11H19NO4 | |
| 1 g | |
| LIWFYAVKYUQMRE-UHFFFAOYSA-N | |
| 1-O-tert-butyl 3-O-methyl pyrrolidine-1,3-dicarboxylate | |
| 4572775 | |
| 97% |
| 105°C (0.2 mmHg) | |
| MFCD04038683 | |
| methyl 1-boc-3-pyrrolidinecarboxylate, 1-tert-butyl 3-methyl pyrrolidine-1,3-dicarboxylate, methyl 1-n-boc-dl-beta-prolinate, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, methyl 1-boc-3-pyrrolidine carboxylate, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, methyl 1-boc-pyrrolidine-3-carboxylate, methyl 1-tert-butoxycarbonyl-3-pyrrolidinecarboxylate, s-1-boc-pyrrolidine-1,3-dicarboxylate, pubchem10748 | |
| CC(C)(C)OC(=O)N1CCC(C1)C(=O)OC | |
| 229.276 | |
| 229.28 | |
| Methyl 1-Boc-pyrrolidine-3-carboxylate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only