missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-Boc-pyrrolidine-3-carboxylate, 97%

Catalog No. AAH3340703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3340703 1 g
AAH3340706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3340703 Supplier Thermo Scientific Chemicals Supplier No. H3340703

CAS: 122684-33-7 | C11H19NO4 | 229.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122684-33-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD04038683
InChI Key LIWFYAVKYUQMRE-UHFFFAOYSA-N
Synonym methyl 1-boc-3-pyrrolidinecarboxylate, 1-tert-butyl 3-methyl pyrrolidine-1,3-dicarboxylate, methyl 1-n-boc-dl-beta-prolinate, 1,3-pyrrolidinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, methyl 1-boc-3-pyrrolidine carboxylate, pyrrolidine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, methyl 1-boc-pyrrolidine-3-carboxylate, methyl 1-tert-butoxycarbonyl-3-pyrrolidinecarboxylate, s-1-boc-pyrrolidine-1,3-dicarboxylate, pubchem10748
PubChem CID 4572775
IUPAC Name 1-O-tert-butyl 3-O-methyl pyrrolidine-1,3-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)C(=O)OC

Specifications

Boiling Point 105°C (0.2 mmHg)
Quantity 1 g
Formula Weight 229.28
Percent Purity 97%
Chemical Name or Material Methyl 1-Boc-pyrrolidine-3-carboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only