missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1,5-Dimethylpyrazole-3-carboxylate 98.0+%, TCI America™

Catalog No. M28631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28631G 1 g
M28635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28631G Supplier TCI America Supplier No. M28631G
Only null left

Chemical Identifiers

CAS 10250-61-0
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.17
MDL Number MFCD06661312
InChI Key WOBRRDCECRRRGS-UHFFFAOYSA-N
Synonym 1,5-Dimethylpyrazole-3-carboxylic Acid Methyl Ester
PubChem CID 2755339
IUPAC Name methyl 1,5-dimethyl-1H-pyrazole-3-carboxylate
SMILES COC(=O)C1=NN(C)C(C)=C1

Specifications

Melting Point 75°C
Color White-Yellow
Boiling Point 144°C
Quantity 1 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1,5-Dimethylpyrazole-3-carboxylate