missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1,2,4-triazole-3-carboxylate, 98%

Catalog No. p-7027751
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2475006 5 g
AAB2475014 25 g
AAB2475022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2475006 Supplier Thermo Scientific Chemicals Supplier No. B2475006
Only null left

CAS: 4928-88-5 | C4H5N3O2 | 127.103 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4928-88-5
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.103
MDL Number MFCD00135989
InChI Key QMPFMODFBNEYJH-UHFFFAOYSA-N
PubChem CID 2735089
IUPAC Name methyl 1H-1,2,4-triazole-5-carboxylate
SMILES COC(=O)C1=NC=NN1

Specifications

Quantity 5 g
Synonym methyl 1h-1,2,4-triazole-3-carboxylate, methyl 1,2,4-triazole-3-carboxylate, 1h-1,2,4-triazole-3-carboxylic acid methyl ester, methyl 4h-1,2,4-triazole-3-carboxylate, 1,2,4-triazole-3-carboxylic acid methylester, 1h-1,2,4-triazole-3-carboxylic acid, methyl ester, 1h-1,2,4-triazole-5-carboxylic acid methyl ester, methyl1,2,4-triazole-3-carboxylate, acmc-20a2ir, 5-carbomethoxy-1,2,4-triazole
Formula Weight 127.1
Percent Purity 98%
Chemical Name or Material Methyl 1,2,4-triazole-3-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only