missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1,2,3,4-Tetra-O-acetyl-beta-D-glucuronate 96.0+%, TCI America™

Catalog No. p-7132886
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
M18681G 1 g
M18685G 5 g
2 options

Catalog No. M18681G

Supplier: TCI America M18681G

Only null left

Chemical Identifiers

CAS 7355-18-2
Molecular Formula C15H20O11
Molecular Weight (g/mol) 376.314
MDL Number MFCD00069834
InChI Key DPOQCELSZBSZGX-XOBFJNJYSA-N
Synonym 2s,3r,4s,5s,6s-6-methoxycarbonyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, methyl o-tetraacetyl-beta-d-glucopyranuronate, methyl 1,2,3,4-tetra-o-acetyl-beta-d-glucuronate, methyl 1,2,3,4-tetra-o-acetyl-beta-d-glucopyranuronate, methyl 2s,3s,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxane-2-carboxylate, 1,2,3,4-tetra-o-acetyl-beta-d-glucopyranuronic acid methyl ester, methyl 1,2,3,4-tetra-o-acetyl-, a-d-glucopyranuronate, beta-d-glucopyranuronic acid, methyl ester, tetraacetate, methyl 1,2,3,4-tetra-o-acetyl-d-glucuronate, 1,2,3,4-tetra-o-acetyl-d-glucuronide methyl ester
PubChem CID 95087
IUPAC Name methyl (2S,3S,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxane-2-carboxylate
SMILES CC(=O)OC1C(C(OC(C1OC(=O)C)OC(=O)C)C(=O)OC)OC(=O)C

Specifications

Melting Point 179°C
Color White-Yellow
Quantity 1 g
Formula Weight 376.31
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 1,2,3,4-Tetra-O-acetyl-beta-D-glucuronate