missing translation for 'onlineSavingsMsg'
Learn More

Methoxypolyethylene glycol amine, M.W. 5,000

Catalog No. p-7080252
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46924MD 250 mg
AA4692403 1 g
AA4692406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA46924MD Supplier Thermo Scientific Chemicals Supplier No. 046924MD
Only null left

CAS: 80506-64-5 | H2NCH2CH2(OCH2CH2)nOCH3 | 559.69 g/mol

Methoxypolyethylene glycol amine is used in the reduction of immunogenicity and increasing serum half life.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 80506-64-5
Molecular Formula H2NCH2CH2(OCH2CH2)nOCH3
Molecular Weight (g/mol) 559.69
MDL Number MFCD00131599
InChI Key QWCGXANSAOXRFE-UHFFFAOYSA-N
Synonym 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine, mpeg12-nh2, mpeg12-amine, m-dpeg12-amine, m-peg12-amine, ambotzpeg1655, m-dpeg r 12-amine, aminopolyethylene glycol 5'000 monomethyl ether, 3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaheptatriacontane-1-amine
IUPAC Name 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine
SMILES COCCOCCN

Specifications

Color White
Physical Form Granular Solid
Solubility Information Soluble in water,methanol,ethanol,chlorinated solvents and terahydrofuran. Insoluble in ether.
Quantity 250 mg
Chemical Name or Material Methoxypolyethylene glycol amine, M.W. 5,000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only