missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxypolyethylene glycol amine, M.W. 10,000
CAS: 80506-64-5 | H2NCH2CH2(OCH2CH2)nOCH3 | 559.69 g/mol
$114.92 - $1428.14
Chemical Identifiers
| CAS | 80506-64-5 |
|---|---|
| Molecular Formula | H2NCH2CH2(OCH2CH2)nOCH3 |
| Molecular Weight (g/mol) | 559.69 |
| MDL Number | MFCD00131599 |
| InChI Key | QWCGXANSAOXRFE-UHFFFAOYSA-N |
| Synonym | 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine, mpeg12-nh2, mpeg12-amine, m-dpeg12-amine, m-peg12-amine, ambotzpeg1655, m-dpeg r 12-amine, aminopolyethylene glycol 5'000 monomethyl ether, 3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaheptatriacontane-1-amine |
| IUPAC Name | 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine |
| SMILES | COCCOCCN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA46433MD
|
Thermo Scientific Chemicals
046433MD |
250 mg |
Each for $114.92
|
|
|||||
|
AA4643303
|
Thermo Scientific Chemicals
04643303 |
1 g |
Each for $303.63
|
|
|||||
|
AA4643306
|
Thermo Scientific Chemicals
04643306 |
5 g |
Each for $1,428.14
|
|
|||||
Description
It is used as a reagent for pegylation of biomolecules, improvement of solubility and other pharmacological parameters, for the preparation of various conjugates containing polyethylene glycol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 80506-64-5 | |
| 559.69 | |
| QWCGXANSAOXRFE-UHFFFAOYSA-N | |
| 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine |
| H2NCH2CH2(OCH2CH2)nOCH3 | |
| MFCD00131599 | |
| 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine, mpeg12-nh2, mpeg12-amine, m-dpeg12-amine, m-peg12-amine, ambotzpeg1655, m-dpeg r 12-amine, aminopolyethylene glycol 5'000 monomethyl ether, 3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaheptatriacontane-1-amine | |
| COCCOCCN |
Specifications
| White | |
| 80506-64-5 | |
| MFCD00131599 | |
| Soluble in water. | |
| QWCGXANSAOXRFE-UHFFFAOYSA-N | |
| 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine | |
| Methoxypolyethylene glycol amine, M.W. 13,000 |
| Granular Solid | |
| H2NCH2CH2(OCH2CH2)nOCH3 | |
| 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine, mpeg12-nh2, mpeg12-amine, m-dpeg12-amine, m-peg12-amine, ambotzpeg1655, m-dpeg r 12-amine, aminopolyethylene glycol 5'000 monomethyl ether, 3,6,9,12,15,18,21,24,27,30,33,36-dodecaoxaheptatriacontane-1-amine | |
| 250 mg | |
| COCCOCCN | |
| 559.69 |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only