missing translation for 'onlineSavingsMsg'
Learn More

Methoxypolyethylene glycol, 5,000 Propionic Acid N-succinimidyl ester, ≥80%, MilliporeSigma™ Supelco™

$1974.70 - $1974.70

Specifications

MDL Number MFCD01867749
Synonym mono-Methyl polyethylene glycol 5'000 propionic acid N-succinimidyl ester; PEG-succinimidyl propionate; Polyethylene glycol N-succinimidyl propionate
Percent Purity ≥80%
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111003110
View Documents
Merck Emd Millipore
859691G
1 g
Each for $1,974.70
Only null left
 
Description

Description

Specifications

Specifications

MFCD01867749
mono-Methyl polyethylene glycol 5'000 propionic acid N-succinimidyl ester; PEG-succinimidyl propionate; Polyethylene glycol N-succinimidyl propionate
≥80%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'