missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxypolyethylene glycol, 5,000 acetic acid N-succinimidyl ester, ≥80%, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 92451-01-9 |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Percent Purity | ≥80% (NMR); ≥80% |