missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxycyclohexane 98.0+%, TCI America™
$296.48 - $995.31
Chemical Identifiers
| CAS | 931-56-6 |
|---|---|
| Molecular Formula | C7H14O |
| Molecular Weight (g/mol) | 114.188 |
| MDL Number | MFCD00154087 |
| InChI Key | GHDIHPNJQVDFBL-UHFFFAOYSA-N |
| PubChem CID | 13607 |
| IUPAC Name | methoxycyclohexane |
| SMILES | COC1CCCCC1 |
Chemical Identifiers
| 931-56-6 | |
| 114.188 | |
| GHDIHPNJQVDFBL-UHFFFAOYSA-N | |
| methoxycyclohexane |
| C7H14O | |
| MFCD00154087 | |
| 13607 | |
| COC1CCCCC1 |
Specifications
| 931-56-6 | |
| Colorless | |
| C7H14O | |
| 5 g | |
| GHDIHPNJQVDFBL-UHFFFAOYSA-N | |
| methoxycyclohexane | |
| 13607 | |
| ≥98.0% (GC) | |
| Methoxycyclohexane |
| -74°C | |
| 135°C | |
| MFCD00154087 | |
| 3271 | |
| COC1CCCCC1 | |
| 114.188 | |
| 114.19 | |
| Liquid |
Safety and Handling
TSCA : No