missing translation for 'onlineSavingsMsg'
Learn More

Methoxycarbonylsulfenyl chloride, 95%

Catalog No. AAL0435003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0435003 1 g
AAL0435006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0435003 Supplier Thermo Scientific Chemicals Supplier No. L0435003

CAS: 26555-40-8 | C2H3ClO2S | 126.554 g/mol

Methoxycarbonylsulfenyl chloride is used to prepare 3-[(methoxycarbonyl)dithio]-L-alanine by reacting with L-cysteine. Further, it is used in the synthesis of 3-2 pyridinyldithio propanoic acid hydrazide (PDPH). It serves as an important heterobifunctional crosslinker, which is a crosslinker used to make macromolecule-drug conjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methoxycarbonylsulfenyl chloride is used to prepare 3-[(methoxycarbonyl)dithio]-L-alanine by reacting with L-cysteine. Further, it is used in the synthesis of 3-2 pyridinyldithio propanoic acid hydrazide (PDPH). It serves as an important heterobifunctional crosslinker, which is a crosslinker used to make macromolecule-drug conjugates.

Solubility
Miscible with dichlormethane.

Notes
Moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 26555-40-8
Molecular Formula C2H3ClO2S
Molecular Weight (g/mol) 126.554
MDL Number MFCD00013648
InChI Key TXJXPZVVSLAQOQ-UHFFFAOYSA-N
Synonym methoxycarbonylsulfenyl chloride, methoxycarbonyl sulfenyl chloride, s-chloro o-methyl thiocarbonate, thiocarbonic acid, anhydrosulphide with methyl thiohypochlorite, carbonothioic acid, anhydrosulfide with thiohypochlorous acid, methyl ester, acmc-209gs8, methoxycarbonylsulfenylchloride, chlorothio methoxy oxomethane, methoxy carbonylsulfenyl chloride, methoxy-carbonylsulfenyl chloride
PubChem CID 123417
IUPAC Name methyl chlorosulfanylformate
SMILES COC(=O)SCl

Specifications

Density 1.398
Boiling Point 73°C to 74°C (100 mmHg)
Flash Point 54°C (129°F)
Refractive Index 1.482
Quantity 1 g
UN Number UN2920
Beilstein 1849908
Sensitivity Moisture sensitive
Solubility Information Miscible with dichlormethane.
Formula Weight 126.56
Percent Purity 95%
Chemical Name or Material Methoxycarbonylsulfenyl chloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 247-801-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only